Azido-PEG8-acid
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Azido-dPEG®8-acid
CAS Number:1214319-92-2
:
Formula:C₁₉H₃₇N₃O₁₀
Molecular Weight:467.51 g/mol
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together