(+)-Sparteine
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Pachycarpine, (7R,7aR,14R,14aS)-Dodecahydro-2H,6H-7,14-methanodipyrido[1,2-a:1',2'-e][1,5]diazocine
CAS Number:492-08-0
:
Formula:C₁₅H₂₆N₂
MDL Number:MFCD00869353
Boiling Pt:174 °C/8mmHg
Density:1.02 g/cm³ (25 °C)
Molecular Weight:234.39 g/mol
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together