Order Entry
Ireland
ContactUsLinkComponent
 
Methyl 2,6-difluoro-4-hydroxybenzoate 98%
Catalog # APOSPC909789-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2,6-difluoro-4-hydroxybenzoate 98%
Catalog # APOSPC909789-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :188.128 g/mol
Storage Temperature:Ambient
CAS Number:194938-88-0
Formula:C₈H₆F₂O₃
MDL Number:MFCD11042796