2-(Aminomethyl)-5-(trifluoromethyl)pyridine hydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 5-(Trifluoromethyl)picolinylamine hydrochloride, [5-(Trifluoromethyl)pyridin-2-yl]methylamine hydrochloride, (5-(Trifluoromethyl)pyridin-2-yl)methanamine hydrochloride
CAS Number:871826-12-9
:
Formula:C₇H₇F₃N₂·ClH
MDL Number:MFCD05863878
Molecular Weight:212.6 g/mol
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together