2,6-Difluoro-3-methoxybenzyl alcohol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886498-45-9
:
Formula:C₈H₈F₂O₂
MDL Number:MFCD04115904
Molecular Weight:174.145 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together