3-(5-Fluoropyridin-2-yl)acrylic acid hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:917760-91-9
:
Formula:C₈H₆FNO₂·ClH
MDL Number:MFCD23135525
Molecular Weight:203.598 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together