4,5-Difluoro-2-methoxybenzeneboronic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: (4,5-Difluoro-2-methoxyphenyl)boronic acid, 4,5-Difluoro-2-methoxyphenylboronic acid, 2-Borono-4,5-difluoroanisole
CAS Number:870777-32-5
:
Formula:C₇H₇BF₂O₃
MDL Number:MFCD08275291
Molecular Weight:187.936 g/mol
Melting Pt:175…180 °C
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together