1-Isopropyl-4-nitro-1H-pyrazole 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:97421-21-1
:
Formula:C₆H₉N₃O₂
MDL Number:MFCD10001542
Molecular Weight:155.155 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together