N-Boc-isoindoline-1-carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2-(tert-Butoxycarbonyl)isoindoline-1-carboxylic acid
CAS Number:221352-46-1
:
Formula:C₁₄H₁₇NO₄
MDL Number:MFCD01321007
Molecular Weight:263.289 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together