(3-Methylpyridin-4-yl)methanamine dihydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85127-50-0
:
Formula:C₇H₁₀N₂·2ClH
MDL Number:MFCD09910172
Molecular Weight:195.09 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together