3-Methoxy-2-nitrophenol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3114-61-2
:
Formula:C₇H₇NO₄
MDL Number:MFCD16876801
Molecular Weight:169.135 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together