Order Entry
Ireland
ContactUsLinkComponent
Methyl 3-(3-hydroxyphenyl)propionate 97%
Methyl 3-(3-hydroxyphenyl)propionate 97%
Catalog # APOSOR912911-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 3-(3-hydroxyphenyl)propionate 97%
Catalog # APOSOR912911-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :180.2 g/mol
Storage Temperature:Ambient
CAS Number:61389-68-2
Formula:C₁₀H₁₂O₃
MDL Number:MFCD03788460