2-Isopropoxy-5-methylphenylboronic acid 95%
Catalog # APOSOR907454-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:194.035 g/mol
Storage Temperature:Ambient
CAS Number:480438-71-9
Formula:C₁₀H₁₅BO₃
MDL Number:MFCD04974067