1-(3-Bromophenyl)cyclopropan-1-amine, N-BOC protected 97%
Catalog # APOSOR61242-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
- Environmentally Preferable:
Specifications
About this item
Synonyms: 1-(3-Bromophenyl)-1-[(tert-butoxycarbonyl)amino]cyclopropane, tert-Butyl [1-(3-bromophenyl)cycloprop-1-yl]carbamate, tert-Butyl (1-(3-bromophenyl)cyclopropyl)carbamate
:
:312.202 g/mol
Storage Temperature:Ambient
CAS Number:597563-17-2
Formula:C₁₄H₁₈BrNO₂
MDL Number:MFCD19381711