N3-Methylpyridine-3,4-diamine ≥98%
Catalog # APOSOR510126-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
:
:123.156 g/mol
Storage Temperature:Ambient
CAS Number:1796-73-2
Formula:C₆H₉N₃
MDL Number:MFCD18384267