Order Entry
Ireland
ContactUsLinkComponent
 
2,3-Dichloro-L-phenylalanine 97%
Catalog # APOSOR452046-1G
Supplier:  Apollo Scientific
CAS Number:  
2,3-Dichloro-L-phenylalanine 97%
Catalog # APOSOR452046-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: (S)-2-Amino-3-(2,3-dichlorophenyl)propanoic acid, H-Phe(2,3-DiCl)-OH
 :
 :234.079 g/mol
Storage Temperature:Ambient
CAS Number:873429-57-3
Formula:C₉H₉Cl₂NO₂
MDL Number:MFCD07372011