Fmoc-3-Bromo-D-b-phenylalanine 97%
Catalog # APOSOR452040-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: (R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-bromophenyl)propanoic acid
:
:466.324 g/mol
Storage Temperature:Ambient
CAS Number:517905-85-0
Formula:C₂₄H₂₀BrNO₄
MDL Number:MFCD03428038