Order Entry
Ireland
ContactUsLinkComponent
 
9,10-Bis(2-naphthyl)anthracene-2-ylboronic acid 95+%
Catalog # APOSOR360482-1G
Supplier:  Apollo Scientific
CAS Number:  
9,10-Bis(2-naphthyl)anthracene-2-ylboronic acid 95+%
Catalog # APOSOR360482-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: (9,10-Di(naphthalen-2-yl)anthracen-2-yl)boronic acid
 :
 :474.356 g/mol
Storage Temperature:Ambient
CAS Number:867044-28-8
Formula:C₃₄H₂₃BO₂
MDL Number:MFCD11045063