tert-Butyl 1,8-diazaspiro[4.5]decane-1-carboxylate 95+%
Catalog # APOSOR312325-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
- Environmentally Preferable:
Specifications
About this item
Synonyms: 1-Boc-1,8-diaza-spiro[4.5]decane
:
:240.342 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:885279-92-5
Formula:C₁₃H₂₄N₂O₂
MDL Number:MFCD06739037