(3R,4R)-4-Methylthio-3-[(R)-1-((t-butyldimethylsilyl)oxy)ethyl]-2-azetidinone
Catalog # APOSOR310547-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:275.483 g/mol
Storage Temperature:Ambient
CAS Number:82864-31-1
Formula:C₁₂H₂₅NO₂SSi
MDL Number:MFCD27957344