1-Methyl-3-nitro-1H-pyrazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:54210-32-1
:
Formula:C₄H₅N₃O₂
MDL Number:MFCD03444368
Molecular Weight:127.101 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together