5-(Aminomethyl)-2,3-dihydrobenzo[b]furan hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: (2,3-Dihydrobenzofuran-5-yl)methanamine hydrochloride, 5-(Aminomethyl)-2,3-dihydro-1-benzofuran hydrochloride, (2,3-Dihydrobenzo[b]furan-5-yl)methylamine hydrochloride
CAS Number:635309-62-5
:
Formula:C₉H₁₁NO·ClH
MDL Number:MFCD03659695
Molecular Weight:185.651 g/mol
Melting Pt:249…250 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together