4-Chloro-4'-iodobiphenyl
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:60200-91-1
:
Formula:C₁₂H₈ClI
MDL Number:MFCD03931233
Molecular Weight:314.549 g/mol
Melting Pt:147…148 °C
:GHS05
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together