Diethylcarbamazine
: 1-(N,N-Diethylcarbamyl)-4-methylpiperazine, N,N-Diethyl-4-methylpiperazine-1-carboxamide
:90-89-1
:MFCD00023288
:C₁₀H₂₁N₃O
:MFCD00039124
:199.293 g/mol
:49…51 °C
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:Almost white to yellow Crystals and chunks
- Infrared spectrum:Conforms
- Melting point:48 to 52 °C
- GC:≥98.5 %
Frequently Bought Together