Thermal transfer printable labels, series R4900
Supplier: Brady
Certificates
About this item
Printable, nylon cloth labels for thermal transfer printers.
- Die-cut labels are on a roll
- Electrical and laboratory application
- Service temperature: –40 to 90 °C
- This R4900 series has matt finish
- Thickness: 0,165 mm
- Material with autoclavable and cryogenic storage properties
This node contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates. This node contains only supplies sold with the ribbon seperate, it is not in the same enclosure as the tape/sleeves.