Wash 96- or 384-well plates securely with the Thermo Scientific™ Wellwash™ Versa Microplate Washer, an easy-to-use microplate-strip washer for routine ELISA applications. Designed for high performance and versatility, enjoy the ease of use and convenience provided through a graphical interface, local language version and USB port.
- Easy and convenient to use
- Large color display and keypad for easy, convenient use
- Clear display shows everything on one screen
- User interface is logic and intuitive, allowing quick protocol development
- Easy-to-use software requires minimal training for fast startup
- Reliable and secure performance
- Unpressurized bottles are safe and secure to use
- Liquid level sensors are present in both wash and waste bottles
- Automatic rinse and prime features assure safe performance
- Aerosol cover prevents aerosolization of infectous diseases
- Plate sensor recognizeds if plate is present
- Optimal washing performance
- Sweep mode guarantees an extremely low residual volume in the well, resulting in reliable assay results
- Washing performance is further optimized by adjustable parameters such as dispense and aspiration height, and aspiration speed
Added versatility-a USB port for transferring protocols between several washers and for printing reports and log files. Specially designed 2x8 cell wash head is available for cell washing. Two 1x8 wash heads, two wash bottles and one rinse bottle, optional two 1x12 wash heads, 1x16 wash heads and 2x8 cell wash head offer increased flexibility. Possibility to connect to automation systems.
Recommended for ELISA-based assays, cell washing.
- Catalog No:
- 736-3009
- 736-0420
- Model:
- Wellwash Versa
- Wellwash Versa
- Plate types:
- 96- and 384-well
- 96- and 384-well
- Wash heads:
- 2×12 cell wash heads
- 2×8 cell wash heads
- Washing volume:
- 50 - 1000 µl (96-well plates)
20 - 300 µl (384-well plates) - 50 - 1000 µl (96-well plates)
20 - 300 µl (384-well plates)
- Dispensing precision:
- <3% (CV) @ 300 µl
- <3% (CV) @ 300 µl
- Dispensing volume range:
- 50 - 400 µl (96-well plates)
20 - 120 µl (384-well plates) - 50 - 400 µl (96-well plates)
20 - 120 µl (384-well plates)
- Display:
- High-contrast colour display with 480×272 dots
- High-contrast colour display with 480×272 dots
- Interfaces:
- USB , RS232C
- USB , RS232C
- Priming and rinsing volume:
- 5 - 100 ml
- 5 - 100 ml
- Residual aspiration volume:
- <1,5 μl
- <1,5 μl
- W×D×H:
- 345×38×240 mm
- 345×38×240 mm
- Wash/waste bottle capacity:
- 2×2 L wash bottles; 1×2 L rinse bottle; 4 L waste bottle
- 2×2 L wash bottles; 1×2 L rinse bottle; 4 L waste bottle
- Weight:
- 9 kg
- 9 kg