Order Entry
Ireland
ContactUsLinkComponent
 
Tetradecyltrimethylammonium bromide for synthesis, Sigma-Aldrich®
 :  Merck
 
Tetradecyltrimethylammonium bromide for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Environmentally Preferable:

 

 

  • Solubility:
    100 g/l (20 °C)
  • Melting Point:
    245 - 250 °C
  • Molar Mass:
    336.4 g/mol
  • Bulk Density:
    600 kg/m3

 

Frequently Bought Together