4-Chloro-3-(trifluoromethyl)phenyl isocyanate 98%
: 4-Chloro-3-(trifluoromethyl)phenylisocyanate, 1-Chloro-4-isocyanato-2-(trifluoromethyl)benzene
:327-78-6
:
:C₈H₃ClF₃NO
:MFCD00013874
:86…90 °C (14 mmHg)
:221.57 g/mol
:29…37 °C
:99 °C
:2811
:6.1 I
:GHS05
:GHS06
:GHS09
:Ambient
- Environmentally Preferable:
- Appearance:White to yellow Powder and/or chunks or low melting solid
- Infrared spectrum:Conforms
- Melting point:29 to 37 °C
- GC:≥97.5 %
Frequently Bought Together