Sodium diethyldithiocarbamate trihydrate ACS
: (Diethyldithiocarbamato-S,S')sodium trihydrate, Diethyldithiocarbamic acid sodium salt trihydrate, N,N-Diethyldithiocarbamic acid sodium salt trihydrate
:20624-25-3
:
:(C₂H₅)₂NCSSNa·3H₂O
:MFCD00150617
:225.31 g/mol
:95…98.5 °C
:9 III
:12,03443
:205-710-6
:3077
:GHS07
:GHS09
- Environmentally Preferable:
- Appearance:Off-white to light grey Powder or crystals
- Infrared spectrum:Conforms
- Solubility:(in water) Passes test
- Sensitivity:(to Copper) Passes test
- Sodium (Na):30.5 to 32.5 % (as Na2SO4)
Frequently Bought Together