Hydriodic acid 57% (w/w) in aqueous solution unstabilised for analysis, distilled
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Hydroiodic acid, Hydrogen iodide, Hydrogen iodide solution
CAS Number:10034-85-2
:
Formula:HI
MDL Number:MFCD00011347
Density:1.7 g/cm³
Molecular Weight:127.91 g/mol
Storage Temperature:Refrigerator
EINECS:233-109-9
UN:1787
ADR:8 II
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Dark red-brown Solution
- Titration with NaOH:≥55.0 %
Specification Test Results
Frequently Bought Together