Hydrochloric acid solution 3 M in methanol, LiChropur™ for GC derivatisation, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:0.871...0.881 g/cm³ (20 °C)
Molecular Weight:36.46 g/mol
Storage Temperature:Freezer
UN:3286
ADR:3 (6.1, 8) II
:GHS02
:GHS05
:GHS06
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (Color):Colorless
- Appearance (Form):Liquid
- Titration (Method):Alkalimetric
- Titration (Molarity):2.7 - 3.3 mol/l
- Remarks on GC:Suitibility for GC Derivatization Complying
- Density D20/4:0.871 - 0.881
- Proton NMR Spectrum:Conforms to Structure
- Recommended Retest Period:5 Months
Specification Test Results
Frequently Bought Together