Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Assay Buffers
J.T.Baker®, Sheath fluid, 3200/3400
J.T.Baker®, Sheath fluid, 3200/3400
3832.9020
:
Avantor
:
J.T.Baker®, Sheath fluid, 3200/3400
3832.9020
:
Avantor
:
Pk:
20 L
Description:
Sheath fluid, 3200/4000
Environmentally Preferable: