Order Entry
Ireland
ContactUsLinkComponent
Polyethylene analytical standards, for GPC
Polyethylene analytical standards, for GPC
Catalog # 81223-100MG
Supplier:  Merck
CAS Number:  
undefined
Polyethylene analytical standards, for GPC
Catalog # 81223-100MG
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    100 mg
  • Description:
    ∼2010 Mw, Mw/Mn ~1,12
  • Environmentally Preferable:
  • M-Clarity quality level:
    MQ100
Specifications
About this item
Certifications: Exact values can be taken from the accompanying certificate of analysis.
Caution: Limited shelf life, expiry date on the label.