Order Entry
Ireland
ContactUsLinkComponent
Methylene blue for microscopy (C.I. 52015)
Methylene blue for microscopy (C.I. 52015)
Supplier:  VWR Chemicals
Certificates
undefined
Methylene blue for microscopy (C.I. 52015)
Supplier:  VWR Chemicals
Certificates
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Proportion A635/A662:
    0.54 --> 0.62