pH micro electrodes, InLab® Micro Pro ISM® / InLab® Ultra-Micro-ISM® / InLab® Surface Pro-ISM
pH electrodes with MultiPin™ connector.
- ARGENTHAL™ reference system with silver-ion trap
- 3 mol/L KCl electrolyte (except InLab® Ultra-Micro-ISM®: FRISCOLYT-B®
The revolutionary Intelligent Sensor Management system (ISM) offers the user an even greater level of security. The Calibration data is stored on the chip in the sensor and is used automatically when the sensor is connected to the meter. This ensures that the meter always uses the latest calibration data.
With integrated NTC 30 kOhm temperature probe (except InLab® Ultra-Micro-ISM®)
* Without NTC 30 kOhm temperature probe
: Supplied with test certificate.
Frequently Bought Together