Multi management software, APT-COM™ 4
Supplier: Binder
Certificates
About this item
This software provides management, logging, programming, documentation options, and much more. With features ranging from GLP-compliant logging to convenient program creation and management of up to 100 units, the APT-COM™ 4 can be used in all industries and makes complex work processes much easier.
- Central overview of all units in both graphic and table form
- Record management (creating, deleting, archiving)
- Choice of five menu languages: DE, EN, FR, IT, ES
- Graphical/numerical program editor
- Reminders can be set via timer function
- Documentation of measured values
- Export of measured values in CSV and PDF format
BASIC Edition: For simple recording and documentation requirements with up to five networked units.
PROFESSIONAL Edition: Convenient unit and user management which builds on the BASIC edition. Suitable for networking up to 100 units.
GLP Edition: For working under GLP-compliant conditions. Measured values are documented in a tamperproof way in line with the requirements of FDA Regulation 21 CFR 11.