Western Blot Transfer buffer, 10X, methanol-free, Pierce™
Catalog # PIER35045
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:500 mL
- Description:Pierce™ 10X Western blot transfer buffer, methanol-free
- Environmentally Preferable:
Specifications
About this item
Pierce™ Methanol-Free 10X Western Blot Transfer Buffer is a space-saving stock solution for preparing the methanol-free transfer buffer used with cooled, tank (wet) western blotting transfer apparatus.
- Diluted 10-fold in water, the solution is ready to use for electrophoresis
- No packets to open, no powder to dissolve, and no methanol required
- Eliminates the possibility of powder remaining in a packet
- Eliminates time spent waiting for powder to dissolve
- Concentrated stock minimizes bench space needed for solutions
This concentrated Buffer is ready to use without having to weigh and dissolve dry ingredients or to adjust the pH with concentrated acid or base.
The 10X Western Blot Transfer Buffer is ready to use upon dilution to 1X with water, since it does not contain methanol and the addition of methanol is not necessary for use in electrophoresis.