Ultra clear cap strips for qPCR
732-0081
- Colour:Clear
- Description:Ultra clear cap strips, 8 per strip
- Seq:02
- Environmentally Preferable:
- Pk:120
PP flat 8-cap strips designed for qPCR and other fluorescence applications.
- Compatible with all Thermo Scientific™ 96-well PCR plates, as well as standard and low profile tube strips
- Specially formulated ultra clear polypropylene enhances signal detection by minimising autoflourescence and maximising transmission
- Highly polished flat surface provides an optically clear window to deliver maximum and consistent signal transmission, critical for accurate qPCR results