Order Entry
Ireland
ContactUsLinkComponent
ToxinSensor™ Gel clot endotoxin assay kit
ToxinSensor™ Gel clot endotoxin assay kit
Catalog # GENSL00351
Supplier:  Genscript
CAS Number:  
ToxinSensor™ Gel clot endotoxin assay kit
Catalog # GENSL00351
Supplier:  Genscript
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Gel Clot Endotoxin Assay Kit
  • Environmentally Preferable:
Specifications
About this item
ToxinSensor™ Gel Clot Endotoxin Assay Kit is intended as an in vitro end-product endotoxin test for human and animal parenteral drugs, biological products, and medical devices. The limulus amebocyte lysate (LAL) test is a qualitative test for Gram-negative bacterial endotoxin.
  • Good reproducibility
  • Ready to use reagents and materials, such as tips, endotoxin-free tubes, etc.
Note: For specimen preparation, the specimen should be certified free of β-glucans contaminant which may come from yeast and cellulosic materials, such as blood products.
Delivery: Kit contains LAL reagent water 10 ml (4 bottles), limulus amebocyte lysate (LAL) 4 ml (2 vials), E. coli endotoxin standard 0.5 EU (2 vials), endotoxin-free vials (5×16 vials), endotoxin-free tips 200 µl (1 box of 96 tips), endotoxin-free tips 1000 µl (2 bags of 12 tips) and incubation rack (1 piece). The kit can be stored at 2 to 8 °C for up to 24 months.