Order Entry
Ireland
ContactUsLinkComponent
Density gradient medium, OptiPrep™
Density gradient medium, OptiPrep™
Catalog # D1556-250ML
undefined
Density gradient medium, OptiPrep™
Catalog # D1556-250ML
Specifications
  • Description:
    OptiPrep™ density gradient medium
  • Environmentally Preferable:
  • Pk:
    250 mL
Specifications
About this item
Density gradient medium is suitable for cell and subcellular organelle isolation, virus purification, and isolation of lipoproteins and macromolecules.
OptiPrep™ Density Gradient Medium has been used in the non enveloped virion isolation from bronchoalveolar lavage.