Hexamethylenetetramine 99+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Hexamethylenetetramine, Hexamine, Urotropine, Formin
CAS Number:100-97-0
:
Formula:C₆H₁₂N₄
MDL Number:MFCD00006895
Density:1.33
Molecular Weight:140.19 g/mol
Melting Pt:280 °C (subl.)
Flash Pt:250 °C (482 °F)
ADR:4.1 III
Merck Index:14,5966
EINECS:202-905-8
UN:1328
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together