Iridium(IV) oxide ≥99.99% (metals basis) Ir ≥ 84.5%, Premion®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Iridium dioxide, Iridium(IV) oxide
CAS Number:12030-49-8
:
Formula:IrO₂
MDL Number:MFCD00011065
Boiling Pt:–27 °C (1013 hPa)
Melting Pt:–102 °C
Density:11.7
Molecular Weight:224.22 g/mol
Storage Temperature:Ambient
EINECS:234-743-9
UN:1479
ADR:5.1 II
:GHS03
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together