1-Naphthyl isocyanate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1-Naphthylisocyanate
CAS Number:86-84-0
:
Formula:C₁₁H₇NO
MDL Number:MFCD00003881
Boiling Pt:269 °C
Melting Pt:4 °C
Density:1.177
Molecular Weight:169.18 g/mol
Flash Pt:135 °C (275 °F)
Storage Temperature:Refrigerator
UN:2206
ADR:6.1 III
Merck Index:12,06497
EINECS:201-703-7
:GHS06
:GHS08
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together