Acetylcholine chloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (2-Acetoxyethyl)trimethylammonium chloride, 2-(Acetyloxy)-N,N,N-trimethylethanaminium chloride, 2-Acetoxy-N,N,N-trimethylethanaminium chloride
CAS Number:60-31-1
:
Formula:H₃CC(O)O(CH₂)₂N(CH₃)₃Cl
MDL Number:MFCD00011698
Molecular Weight:181.66 g/mol
Melting Pt:149…152 °C
Merck Index:13,00089
EINECS:200-468-8
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:146 to 151 °C
- Titration Argentometric:≥98.5 % (on dried basis)
- Loss on drying:≤1.0 % (105 °C, 3 hrs)
- Sulfated ash:≤0.2 %
- pH:5.2 to 9.2 (5 % in water)
- Titration after Ion exchange:≥98.5 %
- Chloride (Cl):≤0.02 %
- Arsenic (As):≤0.0001 %
- Calcium (Ca):≤0.02 %
- Iron (Fe):≤0.001 %
- Magnesium (Mg):≤0.01 %
- Heavy metals (as Pb):≤0.001 %
Specification Test Results
Frequently Bought Together