Cetrimonium bromide 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide
CAS Number:57-09-0
:
Formula:CH₃(CH₂)₁₅N(Br)(CH₃)₃
MDL Number:MFCD00011772
Molecular Weight:364.45 g/mol
Melting Pt:230 °C
Flash Pt:244 °C
ADR:9 III
Merck Index:13,02034
EINECS:200-311-3
UN:3077
:GHS05
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥99.0 %
Specification Test Results
Frequently Bought Together