Order Entry
Ireland
ContactUsLinkComponent
 
(3-Aminopropyl)trimethoxysilane 95%
 
undefined
(3-Aminopropyl)trimethoxysilane 95%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4220 to 1.4250 (20°C, 589 nm)
  • GC:
    ≥94 %

 

 

Frequently Bought Together