Lead(II) oxide, Supelco®
: Merck
: Lead monoxide
:1317-36-8
:
:PbO
:MFCD00011164
:1470
:888
:9.5 g/cm³ (20 °C)
:223.2 g/mol
:215-267-0
:3077
:9 III
:13,05431
:M-Clarity™ quality level = MQ200
:GHS08
:GHS07
:GHS09
- Environmentally Preferable:
- Solubility:0.017 g/l (20 °C)
- Melting Point:888 °C
- Molar Mass:223.19 g/mol
- Bulk Density:3500 - 3700 kg/m3
- Boiling Point:1470 °C
- Density:9.53 g/cm3 (20 °C)
- pH:8 - 9 (100 g/l, H2O, 20 °C) (slurry)
Frequently Bought Together