Aluminium oxide, high surface area catalyst support
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide
CAS Number:1344-28-1
:11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424
Boiling Pt:2980
Melting Pt:2045
Density:3.94 g/cm³ (20 °C)
Molecular Weight:101.96 g/mol
Merck Index:13,00355
Storage Temperature:Ambient
EINECS:215-691-6
REACH:01-2119529248-35
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together