Potassium dihydrogen phosphate 99+%, extra pure
: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate
:7778-77-0
:
:KH₂PO₄
:MFCD00011401
:100
:136.09 g/mol
:253 °C
:13,07744
:Ambient
:231-913-4
:01-2119490224-41
- Environmentally Preferable:
- Appearance:Colorless to white Crystalline powder or crystals
- Loss on drying:≤1.0 % (105°C)
- pH:4.2 to 4.7 (1 % at 20°C)
- Titration with NaOH:≥99.0 % (on dried substance)
- Arsenic (As):≤3 ppm
- Lead (Pb):≤5 ppm
Frequently Bought Together