Cyclohexylméthanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Hexahydrobenzyl alcohol, Cyclohexylmethanol, Cyclohexanemethanol
CAS Number:100-49-2
:
Formula:C₇H₁₄O
MDL Number:MFCD00001510
Boiling Pt:181
Melting Pt:–43 °C
Density:0.926 g/cm³ (20 °C)
Molecular Weight:114.19 g/mol
Merck Index:13,02760
:M-Clarity™ quality level = MQ200
EINECS:202-857-8
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Molar Mass:114.18 g/mol
- Boiling Point:180 - 181 °C
- Flash Point:71 °C
- Density:0.93 g/cm3 (20 °C)
- Hill Formula:C7H14O
Specification Test Results
Frequently Bought Together